Learn More
BIOHIPPO Rat CNP Microsample
SDP
Catalog No. 5000315818
Encompass
Rat CNP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CNP in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 13 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CNP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CNP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CNP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15818 Supplier BIOHIPPO Supplier No. BHE15210743
Rat CNP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CNP in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 13 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CNP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CNP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CNP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color