Learn More
Sino Biological Anti-CNTF Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80012MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CNTF/Ciliary Neurotrophic Factor (rR CNTF/Ciliary Neurotrophic Factor; Catalog#80012-RNAE; P20294; Met1-Met200). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8968