Learn More
Sino Biological Anti-CNTFR Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80019MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat CNTFR (rR CNTFR; Catalog#80019-R08B; Q08406-1; Met 1-Pro 346). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-2873