Learn More
Sino Biological Anti-CNTFR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80019RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CNTFR (Catalog#80019-R08B; Q08406-1; Met1-Pro346). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2872