Learn More
American Research Products Inc Rat Coagulation factor VII F7
Catalog No. 5000328040
Encompass
Rat Coagulation factor VII (F7) ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28040 Supplier American Research Products Inc Supplier No. CSBEL007930RA
Rat Coagulation factor VII (F7) ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma tissue homogenates Method Sandwich