Learn More
American Research Products Inc Rat coagulation factor X FX E
Catalog No. 5000329603
Encompass
Rat coagulation factor X FX ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29603 Supplier American Research Products Inc Supplier No. CSBE08441R
Rat coagulation factor X FX ELISA Kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma Method Sandwich