Learn More
Biomatik Corporation Rat Cofilin 1 (CFL1) CLIA Kit
Supplier:  Biomatik Corporation EKU09668
Encompass_Preferred
Rat Cofilin 1 (CFL1) CLIA Kit for detecting CFL1 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P45592. Gene ID: 29271. Target Synonyms: CFL; p18; N-Cofilin; 18 kDa phosphoprotein; Cofilin 1, Non Muscle. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7180