Learn More
Biomatik Corporation Rat Cofilin 1 (CFL1) ELISA Kit
Supplier:  Biomatik Corporation EKU03290
Encompass_Preferred
Rat Cofilin 1 (CFL1) ELISA Kit for detecting CFL1 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P45592. Gene ID: 29271. Target Synonyms: CFL; p18; N-Cofilin; 18 kDa phosphoprotein; Cofilin 1, Non Muscle. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9049