Learn More
Biomatik Corporation Rat COL1 (Collagen Type Ⅰ) ELISA Kit
Supplier:  Biomatik Corporation EKF58839
Encompass_Preferred
Rat COL1 (Collagen Type Ⅰ) ELISA Kit for detecting COL1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.094ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P02454. Target Synonyms: COL1; Alpha-1 type I collagen; Collagen alpha-1; I chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6980