Learn More
Sino Biological Rat COL4A3 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80439R01H100
Encompass_Preferred
A DNA sequence encoding the rat COL4A3 (NP_001129231) (Gly1426-His1670) was expressed with Fc region of human IgG1 at the N-terminus.
Catalog No. 50-237-3797