Learn More
American Research Products Inc Rat Collagen alpha 1 I chain
Catalog No. 5000329791
Encompass
Rat Collagen alpha-1 (I) chain (COL1A1) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29791 Supplier American Research Products Inc Supplier No. CSBE12732R
Rat Collagen alpha-1 (I) chain (COL1A1) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich