Learn More
Biomatik Corporation Rat Collagen Type II Alpha 1 (COL2a1) ELISA Kit
Supplier:  Biomatik Corporation EKU03313
Encompass_Preferred
Rat Collagen Type II Alpha 1 (COL2a1) ELISA Kit for detecting COL2a1 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 46.88-3000pg/ml. Sensitivity: 18.25pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05539. Gene ID: 25412. Target Synonyms: COL2-A1; COL2A1; SEDC; Chondrocalcin; Collagen Alpha-1 (II) Chain; Primary Osteoarthritis,Spondyloepiphyseal Dysplasia,Congenital. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9046