Learn More
Biomatik Corporation Rat Complement 1 Inhibitor (C1INH) ELISA Kit
Supplier:  Biomatik Corporation EKU10739
Encompass_Preferred
Rat Complement 1 Inhibitor (C1INH) ELISA Kit for detecting C1INH in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 6.25-400ng/ml. Sensitivity: 2.61ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6P734. Gene ID: 295703. Target Synonyms: SERPING1; C1IN; C1-INH; C1NH; HAE1; HAE2; Serpin Peptidase Inhibitor Clade G Member 1; Angioedema Hereditary; C1 Inhibitor; C1 Esterase Inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8300