Learn More
Crystal Chem Inc Rat Complement 3 (C3) ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80667
Encompass
The Rat Complement 3 ELISA is used to quantify levels of rat complement 3 (C3) in serum and plasma using only a 5 µL sample volume. Highlights: Kit uses sample size of only 5 µL. High sensitivity of 4.4 ng/mL. Dynamic range of 12.5 - 800 ng/mL. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7796