Learn More
Abcam Rat Complement C3 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1577311X96TESTS
Encompass_Preferred
Rat Complement C3 ELISA Kit ab157731 is used to quantify Rat Complement C3 with a sensitivity of 15.8 pg/ml. Sandwich ELISA Kit
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1539743