Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat Complement C4 ELISA Kit

Catalog No. EEL225
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL225 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL225 Supplier Invitrogen™ Supplier No. EEL225
Add to Cart
Add to Cart

ELISA

Rat Complement C4 ELISA Kit quantifies Complement C4 in serum and plasma. The Rat Complement C4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation. For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

This gene encodes the acidic form of complement factor 4, part of the classical activation pathway. The protein is expressed as a single chain precursor which is proteolytically cleaved into a trimer of alpha, beta, and gamma chains prior to secretion. The trimer provides a surface for interaction between the antigen-antibody complex and other complement components. The alpha chain may be cleaved to release C4 anaphylatoxin, a mediator of local inflammation. Deficiency of this protein is associated with systemic lupus erythematosus and type I diabetes mellitus. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6. Varying haplotypes of this gene cluster exist, such that individuals may have 1, 2, or 3 copies of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08649
Assay Range 3.13-200 ng/mL
Assay Sensitivity 1.88 ng/mL
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum
Target Species Rat
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias acidic C4; acidic complement C4; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; C4; c4 complement; C4-1; C4a; C4a anaphylatoxin; C4A2; C4A3; C4A4; C4A6; C4AD; C4B; C4B_2; C4B1; C4B12; C4B2; C4B3; C4B5; C4b-A; C4b-B; C4BD; C4d-A; C4d-B; C4F; C4S; CH; Chido form of C4; CO4; Complement C4; Complement C4 alpha chain; Complement C4 beta chain; Complement C4 gamma chain; Complement C4-A; Complement C4-A alpha chain; Complement C4-B; Complement C4-B alpha chain; complement C4B1a; complement component 4 (within H-2S); complement component 4, gene 1; complement component 4A (Rodgers blood group); complement component 4B (Chido blood group); complement component 4B (Childo blood group); CPAMD2; CPAMD3; DADB-112B14.11; DAMC-258G8.6; FLJ60561; MGC164979; MHC class III region complement; RG; Rodgers form of C4; sex-limited protein; Slp; Ssacidic C4; acidic complement C4; Basic complement C4; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 2; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 3; C4; c4 complement; C4-1; C4a; C4a anaphylatoxin; C4A2; C4A3; C4A4; C4A6; C4AD; C4B; C4B_2; C4B1; C4B12; C4B2; C4B3; C4B5; C4b-A; C4b-B; C4BD; C4d-A; C4d-B; C4F; C4S; CH; Chido form of C4; CO4; Complement C4; Complement C4 alpha chain; Complement C4 beta chain; Complement C4 gamma chain; Complement C4-A; Complement C4-A alpha chain; Complement C4-B; Complement C4-B alpha chain; complement C4B1a; complement component 4 (within H-2S); complement component 4, gene 1; complement component 4A (Rodgers blood group); complement component 4B (Chido blood group); complement component 4B (Childo blood group); CPAMD2; CPAMD3; DADB-112B14.11; DAMC-258G8.6; FLJ60561; MGC164979; MHC class III region complement; RG; Rodgers form of C4; sex-limited protein; Slp; Ss;
Gene ID (Entrez) 24233, 406161
Gene Symbol C4A, C4B
Interassay CV <10%
Intraassay CV <10%
Kit Contents Micro ELISA Plate (Dismountable), Reference Standard , Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis, Reference Standard, Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma: 100 μL, Serum: 100 μL
Shipping Condition Wet Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Complement C4
Test Time 1 hr 20 min
Total Assay Time 3 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.