Learn More
Biomatik Corporation Rat Complement Component 1, Q Subcomponent B (C1qB) ELISA Kit
Supplier:  Biomatik Corporation EKU09149
Encompass_Preferred
Rat Complement Component 1, Q Subcomponent B (C1qB) ELISA Kit for detecting C1qB in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.64ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31721. Gene ID: 29687. Target Synonyms: Complement Component 1,q Subcomponent,Beta Polypeptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8560