Learn More
Biomatik Corporation Rat Complement Component 5a (C5a) CLIA Kit
Supplier:  Biomatik Corporation EKU09837
Encompass_Preferred
Rat Complement Component 5a (C5a) CLIA Kit for detecting C5a in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.27-200ng/ml. Sensitivity: 0.11ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P08650. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6710