Learn More
American Research Products Inc Rat Complement fragment 5 C5
Catalog No. 5000328643
Encompass
Rat Complement fragment 5 C5 ELISA Kit from CUSABIO Detection Range 9 38 ng/mL-600 ng/mL Sensitivity 2 34 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28643 Supplier American Research Products Inc Supplier No. CSBE14363R
Rat Complement fragment 5 C5 ELISA Kit from CUSABIO Detection Range 9 38 ng/mL-600 ng/mL Sensitivity 2 34 ng/mL Sample serum plasma tissue homogenates Method Sandwich