Learn More
American Research Products Inc Rat connectiVe tissue growth f
Catalog No. 5000326491
Encompass
Rat connectiVe tissue growth factor CTGF ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26491 Supplier American Research Products Inc Supplier No. CSBE07876R
Rat connectiVe tissue growth factor CTGF ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich