Learn More
Biomatik Corporation Rat Coronin 1A (CORO1A) ELISA Kit
Supplier:  Biomatik Corporation EKU03461
Encompass_Preferred
Rat Coronin 1A (CORO1A) ELISA Kit for detecting CORO1A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91ZN1. Gene ID: 155151. Target Synonyms: CLABP; p57; CLIPINA; HCORO1; TACO; Coronin-1; Coronin,Actin-Binding Protein,1A; Coronin-like protein A; Coronin-like protein p57; Tryptophan aspartate-containing coat protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9035