Learn More
Biomatik Corporation Rat Corticotropin Releasing Hormone Binding Protein (CRHBP) ELISA Kit
Supplier:  Biomatik Corporation EKU03471
Encompass_Preferred
Rat Corticotropin Releasing Hormone Binding Protein (CRHBP) ELISA Kit for detecting CRHBP in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24388. Target Synonyms: CRF-BP; CRFBP; Corticotropin-Releasing Factor-Binding Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5565