Learn More
Biomatik Corporation Rat Corticotropin Releasing Hormone Receptor 1 (CRHR1) ELISA Kit
Supplier:  Biomatik Corporation EKU03473
Encompass_Preferred
Rat Corticotropin Releasing Hormone Receptor 1 (CRHR1) ELISA Kit for detecting CRHR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.128ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35353. Gene ID: 58959. Target Synonyms: CRF-R; CRF1; CRFR1; CRH-R1h; CRHR; CRHR1f; Corticotropin-releasing factor receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9033