Learn More
Biomatik Corporation Rat Corticotropin Releasing Hormone Receptor 2 (CRHR2) ELISA Kit
Supplier:  Biomatik Corporation EKU03475
Encompass_Preferred
Rat Corticotropin Releasing Hormone Receptor 2 (CRHR2) ELISA Kit for detecting CRHR2 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.134ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D4A5C4. Target Synonyms: CRFR2; CRF2; Corticotropin-releasing factor receptor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4580