Learn More
Biomatik Corporation Rat Cortisol Elisa Kit
Supplier:  Biomatik Corporation EKC31485
Encompass_Preferred
Rat Cortisol Elisa Kit for detecting Cortisol in serum, plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.049-200ng/ml. Sensitivity: 0.049ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7399