Learn More
Biomatik Corporation Rat Coupling Factor 6 (CF6) ELISA Kit
Supplier:  Biomatik Corporation EKU03480
Encompass_Preferred
Rat Coupling Factor 6 (CF6) ELISA Kit for detecting CF6 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21571. Gene ID: 94271. Target Synonyms: ATP5J; ATP5A; ATP5; ATPM; ATP Synthase,H+ Transporting,Mitochondrial Fo Complex,Subunit F6; ATP synthase-coupling factor 6, mitochondrial. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4736