Learn More
Sino Biological Anti-Carboxypeptidase A2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80715RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Carboxypeptidase A2 / CPA2 (rR Carboxypeptidase A2 / CPA2; Catalog#80715-R08H; NP_001013101.2; Met1-Tyr417). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0862