Learn More
BIOHIPPO Rat CR Microsample
SDP
Catalog No. 5000315358
Encompass
Rat CR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CR in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 79-50 ng/mL with a sensitivity of 0 26 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15358 Supplier BIOHIPPO Supplier No. BHE15210601
Rat CR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CR in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 79-50 ng/mL with a sensitivity of 0 26 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color