Learn More
American Research Products Inc Rat Creatine Kinase MB isoenzy
Catalog No. 5000329342
Encompass
Rat Creatine Kinase MB isoenzyme CK-MB ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum urine tissue homogenates plasma Method Sandwich
Catalog No. 50-003-29342 Supplier American Research Products Inc Supplier No. CSBE14403R
Rat Creatine Kinase MB isoenzyme CK-MB ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum urine tissue homogenates plasma Method Sandwich