Learn More
American Research Products Inc Rat Creatine Kinase MM isoenzy
Catalog No. 5000329343
Encompass
Rat Creatine Kinase MM isoenzyme CK-MM ELISA Kit from CUSABIO Detection Range 7 8 U/mL-500 U/mL Sensitivity 1 95 U/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29343 Supplier American Research Products Inc Supplier No. CSBE14405R
Rat Creatine Kinase MM isoenzyme CK-MM ELISA Kit from CUSABIO Detection Range 7 8 U/mL-500 U/mL Sensitivity 1 95 U/mL Sample serum plasma tissue homogenates Method Sandwich