Learn More
Biomatik Corporation Rat Creatine Transporter (CRT) ELISA Kit
Supplier:  Biomatik Corporation EKU12227
Encompass_Preferred
Rat Creatine Transporter (CRT) ELISA Kit for detecting CRT in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 246.9-20,000pg/ml. Sensitivity: 105.8pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P28570. Gene ID: 50690. Target Synonyms: SLC6A8; CT1; CRTR; Sodium And Chloride Dependent Creatine Transporter 1; Solute Carrier Family 6 Member 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4393