Learn More
Crystal Chem Inc Rat Creatinine Assay Kit, 5-Kit Pack, 96-tests per kit
SDP
Catalog No. 501947692
Encompass_Preferred
The Rat Creatinine Assay is used to quantify levels of creatinine in rat serum and urine in less than 10 min. The 5-Kit Pack is convieniently provided as five individual kits.Highlights: Fully enzymatic method with significantly reduced interference compared to alkaline picric acid. Less than 10% variance from triglyceride, ascorbic acid, bilirubin, hemoglobinRequires only eight (8) µL of sample, ideal for research applications. Designed for rat samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7692 Supplier Crystal Chem Inc Supplier No. 80345
The Rat Creatinine Assay is used to quantify levels of creatinine in rat serum and urine in less than 10 min. The 5-Kit Pack is convieniently provided as five individual kits.Highlights: Fully enzymatic method with significantly reduced interference compared to alkaline picric acid. Less than 10% variance from triglyceride, ascorbic acid, bilirubin, hemoglobinRequires only eight (8) µL of sample, ideal for research applications. Designed for rat samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.