Learn More
Sino Biological Anti-C-Reactive Protein Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80041T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat C-Reactive Protein/CRP (rR C-Reactive Protein/CRP; Catalog#80041-R08H; NP_058792.1; Met1-Ser230). C-Reactive Protein/CRP specific IgG was purified by Rat C-Reactive Protein/CRP affinity chromatography.
Catalog No. 50-241-0983