Learn More
Sino Biological Anti-C-Reactive Protein Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80041RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CRP / C-Reactive Protein (rR CRP / C-Reactive Protein; Catalog#80041-R08H; NP_058792.1; Met 1-Ser 230). CRP / C-Reactive Protein specific IgG was purified by Rat CRP / C-Reactive Protein affinity chromatography.
Catalog No. 50-240-7871