Learn More
Sino Biological Rat CSF1R PAb 100ul (Sino Biological)
Supplier:  Sino Biological 80447RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CSF1R / MCSF Receptor / CD115 (rR CSF1R / MCSF Receptor / CD115; Catalog#80447-R08H; D4ACA7; Met1-Glu510). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-2516