Learn More
Sino Biological Anti-GM-CSF/CSF2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80020T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat GM-CSF / CSF2 (Catalog#80020-R07H; P48750; Ala18-Lys144). GM-CSF / CSF2 specific IgG was purified by Rat GM-CSF / CSF2 affinity chromatography.
Catalog No. 50-240-8623