Learn More
Sino Biological Anti-Cystatin C Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80040T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Cystatin C (rR Cystatin C; Catalog#80040-R08H; P14841-1; Met1-Ala140). Cystatin C specific IgG was purified by Rat Cystatin C affinity chromatography.
Catalog No. 50-255-2852