Learn More
Sino Biological Anti-CTHRC1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80712T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CTHRC1 (rR CTHRC1; Catalog#80712-R08H; NP_001258229.1; Met1-Lys230). CTHRC1 specific IgG was purified by Rat CTHRC1 affinity chromatography.
Catalog No. 50-255-2760