Learn More
Sino Biological Anti-CTLA-4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 81069T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CTLA-4 / CD152 (rR CTLA-4 / CD152; Catalog#81069-R08H; NP_113862.1; Met1-Asp161). CTLA-4 / CD152 specific IgG was purified by Rat CTLA-4 / CD152 affinity chromatography.
Catalog No. 50-241-1832