Learn More
BIOHIPPO Rat CTSD Microsample
SDP
Catalog No. 5000313812
Encompass
Rat CTSD Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CTSD in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 29 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CTSD Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CTSD Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CTSD biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13812 Supplier BIOHIPPO Supplier No. BHE15211158
Rat CTSD Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CTSD in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 29 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CTSD Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CTSD Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CTSD biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color