Learn More
American Research Products Inc Rat CX3C chemokine receptor 1
Catalog No. 5000328113
Encompass
Rat CX3C chemokine receptor 1 (CX3CR1) ELISA kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-28113 Supplier American Research Products Inc Supplier No. CSBEL006236RA
Rat CX3C chemokine receptor 1 (CX3CR1) ELISA kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich