Learn More
Sino Biological Anti-CXCL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80039T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat CXCL1 / MGSA / NAP-3 (Catalog#80039-RNCE; NP_110472.1; Ala25-Lys96). CXCL1 / MGSA / NAP-3 specific IgG was purified by Rat CXCL1 / MGSA / NAP-3 affinity chromatography.
Catalog No. 50-255-2853