Learn More
Abcam Rat CXCL5 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1007741X96TESTS
Encompass_Preferred
Rat CXCL5 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat CXCL5 in Rat in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2743339