Learn More
BIOHIPPO Rat CXCR3 Microsample
SDP
Catalog No. 5000315240
Encompass
Rat CXCR3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CXCR3 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CXCR3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CXCR3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CXCR3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15240 Supplier BIOHIPPO Supplier No. BHE15212230
Rat CXCR3 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CXCR3 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 06 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat CXCR3 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat CXCR3 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat CXCR3 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color