Learn More
Biomatik Corporation Rat Cyclin Dependent Kinase 2 (CDK2) ELISA Kit
Supplier:  Biomatik Corporation EKU03537
Encompass_Preferred
Rat Cyclin Dependent Kinase 2 (CDK2) ELISA Kit for detecting CDK2 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.133ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q63699. Target Synonyms: CDKN2; p33 (CDK2) ; Cell division protein kinase 2; p33 protein kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9025