Learn More
Biomatik Corporation Rat Cyclin Dependent Kinase Inhibitor 1A (CDKN1A) ELISA Kit
Supplier:  Biomatik Corporation EKU03544
Encompass_Preferred
Rat Cyclin Dependent Kinase Inhibitor 1A (CDKN1A) ELISA Kit for detecting CDKN1A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q64315. Gene ID: 114851. Target Synonyms: WAF1; p21; CIP1; p21WAF1; CAP20; CDKN1; MDA-6; SDI1; p21CIP1; CDK-Interacting Protein 1; Melanoma differentiation-associated protein 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5121