Learn More
Biomatik Corporation Rat Cyclophilin 40 (CYP-40) ELISA Kit
Supplier:  Biomatik Corporation EKU10765
Encompass_Preferred
Rat Cyclophilin 40 (CYP-40) ELISA Kit for detecting CYP-40 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6DGG0. Gene ID: 361967. Target Synonyms: PPID; CYPD; CyP-D; PPIase D; Peptidylprolyl Isomerase D; Cyclophilin 40; 40 kDa peptidyl-prolyl cis-trans isomerase; Cyclophilin-related protein; Rotamase D. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4669