Learn More
Biomatik Corporation Rat Cystatin A (CSTA) ELISA Kit
Supplier:  Biomatik Corporation EKU09002
Encompass_Preferred
Rat Cystatin A (CSTA) ELISA Kit for detecting CSTA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01039. Gene ID: 288075. Target Synonyms: Cys-A; STF1; STFA; Stefin A; Cystatin-AS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5746