Learn More
Biomatik Corporation Rat Cystatin B (CSTB) ELISA Kit
Supplier:  Biomatik Corporation EKU10770
Encompass_Preferred
Rat Cystatin B (CSTB) ELISA Kit for detecting CSTB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.133ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01041. Gene ID: 25308. Target Synonyms: Cys-B; CST6; EPM1; PME; STFB; CPI-B; Stefin B; Liver thiol proteinase inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6679