Learn More
Biomatik Corporation Rat Cystatin C (Cys-C) ELISA Kit
Supplier:  Biomatik Corporation EKU03564
Encompass_Preferred
Rat Cystatin C (Cys-C) ELISA Kit for detecting Cys-C in serum, plasma, urine, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14841. Gene ID: 25307. Target Synonyms: CST3; Cystatin 3; Gamma Trace; Post-Gamma-Globulin; Neuroendocrine Basic Polypeptide; Amyloid Angiopathy And Cerebral Hemorrhage. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7494